Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Ammonium Molybdate, Tetrahydrate
SCI_ED

Catalog No. S25171 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
ACS Grade
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25171 100 g ACS Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25171
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 12027-67-7
Molecular Formula H24Mo7N6O24
Molecular Weight (g/mol) 1163.85
MDL Number MFCD00064633
InChI Key MISTUIVWPMGXFX-UHFFFAOYSA-T
Synonym hexaammonium molybdate, ammoniummolybdatehydrate, nh4 6mo7o4
PubChem CID 71306766
IUPAC Name hexaammonium dodecaoxododecacyclo[7.5.1.1³,⁵.1³,¹³.1⁵,⁷.1⁷,⁹.0¹,⁴.0¹,⁶.0¹,⁸.0²,¹³.0¹¹,¹⁴.0¹¹,¹⁵]heptamolybdoxane-2,4,6,8,14,15-hexakis(ylium)
SMILES [NH4+].[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].O=[Mo]123(=O)O[Mo]45(=O)(=O)O[Mo]67(=O)(=O)O[Mo]89(=O)(=O)O[Mo]%10%11(=O)(=O)O[Mo]%12(=O)(=O)(O1)[O+]2[Mo]([O+]34)([O+]56)([O+]%10%12)([O+]78)[O+]9%11

Specifications

Quantity 100 g
Linear Formula (NH4)6Mo7O24·4H2O
Formula Weight 1235.86
Grade ACS
Packaging ACS Grade
Density 2.49g/mL
Chemical Name or Material Ammonium Molybdate, Tetrahydrate
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.