Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Mucate, MP Biomedicals

Catalog No. ICN205054 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN205054 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN205054 Supplier MP Biomedicals, Inc Supplier No. 0520505480

Chemical Identifiers

CAS 527-04-8
Molecular Formula C6H18N2O8+2
Molecular Weight (g/mol) 246.216
InChI Key LZWIOJIGSVGZID-SXKXKJGMSA-P
Synonym galactaric acid diammonium salt
PubChem CID 101318330
IUPAC Name diazanium;(2S,3R,4S,5R)-2,3,4,5-tetrahydroxyhexanedioic acid
SMILES C(C(C(C(=O)O)O)O)(C(C(=O)O)O)O.[NH4+].[NH4+]

Specifications

Quantity 100 g
Chemical Name or Material Ammonium Mucate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.