Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Ammonium Nitrate
SCI_ED

Catalog No. S25160 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
ACS Grade
Lab Grade
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25160 500 g Lab Grade
S25172 500 g ACS Grade
S25173 100 g Lab Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S25160
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Explosive under high temperatures

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 6484-52-2
Molecular Formula H4N2O3
Molecular Weight (g/mol) 80.04
MDL Number MFCD00011425
InChI Key DVARTQFDIMZBAA-UHFFFAOYSA-O
Synonym ammonium nitrate, nitram, ammonium nitricum, ammonium saltpeter, nitrate of ammonia, nitric acid ammonium salt, nitrato amonico, herco prills, german saltpeter, merco prills
PubChem CID 22985
ChEBI CHEBI:63038
IUPAC Name ammonium nitrate
SMILES [NH4+].[O-][N+]([O-])=O

Specifications

Color White
Physical Form Solid
Quantity 500 g
Assay Percent Range >95% w/v
Linear Formula NH4NO3
Formula Weight 80.04
Packaging Lab Grade
Density 1.72g/mL
Chemical Name or Material Ammonium Nitrate
Safety handling Check Alert storage code Red
ShelfLife 2 years
DOTInformation DOT Class 5.1, III, : Oxidizer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.