Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium nitrate, 98%, ACS reagent, Thermo Scientific Chemicals

Catalog No. AC423350050
Change view
Click to view available options
Quantity:
5 kg
Packaging:
Plastic drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC423350050 5 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC423350050 Supplier Thermo Scientific Chemicals Supplier No. 423350050
  • $414.00
    $338.65 / Each of 1

    Save $75.35

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6484-52-2
Molecular Formula H4N2O3
Molecular Weight (g/mol) 80.04
MDL Number MFCD00011425
InChI Key DVARTQFDIMZBAA-UHFFFAOYSA-O
Synonym ammonium nitrate, nitram, ammonium nitricum, ammonium saltpeter, nitrate of ammonia, nitric acid ammonium salt, nitrato amonico, herco prills, german saltpeter, merco prills
PubChem CID 22985
ChEBI CHEBI:63038
IUPAC Name ammonium nitrate
SMILES [NH4+].[O-][N+]([O-])=O

Specifications

Boiling Point 210.0°C
Melting Point 169.0°C
pH 4.5 to 6.0 (at 5% 25°C)
Quantity 5 kg
Packaging Plastic drum
Residue after Ignition 0.01% max
Specific Gravity 1.72
Assay Percent Range 98%
Density 1.7200g/mL
Linear Formula NH4NO3
Fieser 01,39; 15,14
Merck Index 15,531
Solubility Information Solubility in water: 190 g/100mL (20°C). Other solubilities: 1 g/20mL alcohol, 1 g/8mL methanol
Formula Weight 80.04
Percent Purity 98%
Grade ACS Reagent
Chemical Name or Material Ammonium nitrate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May intensify fire; oxidizer.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Keep/Store away from clothing/ combustible materials.
In case of fire: Use water spray for extinction.
Wear protective gloves/protective clothing/
EINECSNumber 229-347-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.