Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium nitrate, 99+%, for analysis

Catalog No. AC205865000
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC205865000 500 g Plastic bottle
AC205861000 100 g Plastic bottle
AC205860010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC205865000 Supplier Thermo Scientific Chemicals Supplier No. 205865000
Add to Cart

Ammonium nitrate, 99+%, H4N2O3, CAS Number-6484-52-2, ammonium nitricum, ammonium saltpeter, merco prills, herco prills, nitric acid ammonium salt, nitram, nitrato amonico, nitrate of ammonia, german saltpeter, ammonium nitrate, 500g, 210.0 deg.C, CHEBI:63038, 5ppm max, 5ppm max., White, 1.7200g/mL | CAS: 6484-52-2 | H4N2O3 | 80.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6484-52-2
Molecular Formula H4N2O3
Molecular Weight (g/mol) 80.04
MDL Number MFCD00011425
InChI Key DVARTQFDIMZBAA-UHFFFAOYSA-O
Synonym ammonium nitrate, nitram, ammonium nitricum, ammonium saltpeter, nitrate of ammonia, nitric acid ammonium salt, nitrato amonico, herco prills, german saltpeter, merco prills
PubChem CID 22985
ChEBI CHEBI:63038
SMILES [NH4+].[O-][N+]([O-])=O

Specifications

Boiling Point 210.0°C
Melting Point 169.0°C
Color White
pH 4.5 to 6 (5% soln. at 20°C)
Physical Form Adhering Crystals
Quantity 500 g
Assay Percent Range 99+%
Linear Formula NH4NO3
Fieser 01,39; 15,14
Merck Index 15,531
Solubility Information Solubility in water: 190 g/100mL (20°C). Other solubilities: 1 g/20mL alcohol, 1 g/8mL methanol
IUPAC Name ammonium nitrate
Formula Weight 80.04
Percent Purity ≥99%
Grade Analytical
Packaging Plastic bottle
Residue after Ignition 0.01% max. (as SO4)
Specific Gravity 1.72
Density 1.7200g/mL
Chemical Name or Material Ammonium nitrate, p.a.
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May intensify fire; oxidizer.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Keep/Store away from clothing/ combustible materials.
In case of fire: Use water spray for extinction.
Wear protective gloves/protective clothing/
EINECSNumber 229-347-8
RTECSNumber BR9050000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.