Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium nitrate Analytical Reagent, Reag. ACS Reag. Ph. Eur., Solstice

Catalog No. 60045904 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
6 x 1 kg
Packaging:
Plastic Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-904 6 x 1 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-045-904 Supplier Solstice Supplier No. 311146X1KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥98%

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories: Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications. Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Specifications

CAS 6484-52-2
Physical Form Crystalline
Quantity 6 x 1 kg
Packaging Plastic Bottle
Molecular Formula H4N2O3
MDL Number MFCD00011425
InChI Key DVARTQFDIMZBAA-UHFFFAOYSA-O
SMILES [NH4+].[O-][N+]([O-])=O
IUPAC Name ammonium nitrate
Molecular Weight (g/mol) 80.04
PubChem CID 22985
ChEBI CHEBI:63038
Percent Purity ≥98.0%
Grade ACS
Show More Show Less
Recommended Storage Room Temperature
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.