Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Nitrate, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. 02002308 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-002-308 500 g Poly Bottle
02-002-307 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 02-002-308 Supplier Avantor Supplier No. 072901

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 6484-52-2
Molecular Formula H4N2O3
Molecular Weight (g/mol) 80.04
MDL Number MFCD00011425
InChI Key DVARTQFDIMZBAA-UHFFFAOYSA-O
Synonym ammonium nitrate, nitram, ammonium nitricum, ammonium saltpeter, nitrate of ammonia, nitric acid ammonium salt, nitrato amonico, herco prills, german saltpeter, merco prills
PubChem CID 22985
ChEBI CHEBI:63038
IUPAC Name ammonium nitrate
SMILES [NH4+].[O-][N+]([O-])=O

Specifications

Quantity 500 g
Formula Weight 80.04
Grade Meets ACS reagent chemical requirements
Packaging Poly Bottle
Specific Gravity 1L = 1.73kg
Chemical Name or Material Ammonium Nitrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.