Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Nitrate, MilliporeSigma™

Catalog No. M1011885000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1011885000 5 kg Plastic Bottle
M1011880500 500 g Plastic Bottle
M1011881000 1 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1011885000 Supplier MilliporeSigma Supplier No. 1011885000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For analysis EMSURE™ ACS

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6484-52-2
Molecular Formula H4N2O3
Molecular Weight (g/mol) 80.04
MDL Number MFCD00011425
InChI Key DVARTQFDIMZBAA-UHFFFAOYSA-O
Synonym ammonium nitrate, nitram, ammonium nitricum, ammonium saltpeter, nitrate of ammonia, nitric acid ammonium salt, nitrato amonico, herco prills, german saltpeter, merco prills
PubChem CID 22985
ChEBI CHEBI:63038
IUPAC Name ammonium nitrate
SMILES [NH4+].[O-][N+]([O-])=O

Specifications

Boiling Point 210°C
Physical Form Solid
Quantity 5 kg
UN Number UN1942
Solubility Information Soluble in water: 1920g/L at 20°C
Formula Weight 80.04
Percent Purity ≥99.7%
Grade ACS
Odor Odorless
Packaging Plastic Bottle
Density 1.72g/cm3 at 20°C
Chemical Name or Material Ammonium nitrate
Show More Show Less
Hazard Category H272: May intensify fire; oxidiser.
DOTInformation DOT Class: 5.1, Packaging Group: III
EINECSNumber 229-347-8
RTECSNumber BR9050000
Recommended Storage Store at 5° to 30°C (41° to 86°F), Tightly closed. Dry. Protect from Light. Away from combustible material and sources of ignition and heat.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.