Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Nitrate, Extra Pure, MilliporeSigma™

Catalog No. M1011871000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1011871000 1 kg Plastic Bottle
M1011875000 5 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1011871000 Supplier MilliporeSigma Supplier No. 1.01187.1000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recommended Storage

5°C to +30°C

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6484-52-2
Molecular Formula H4N2O3
Molecular Weight (g/mol) 80.04
MDL Number MFCD00011425
InChI Key DVARTQFDIMZBAA-UHFFFAOYSA-O
Synonym ammonium nitrate, nitram, ammonium nitricum, ammonium saltpeter, nitrate of ammonia, nitric acid ammonium salt, nitrato amonico, herco prills, german saltpeter, merco prills
PubChem CID 22985
ChEBI CHEBI:63038
IUPAC Name ammonium nitrate
SMILES [NH4+].[O-][N+]([O-])=O

Specifications

Boiling Point 210°C (15.0hPa)
Melting Point 169°C
pH 4.5 to 7.0
Physical Form Solid
Quantity 1 kg
Solubility Information 1920g/L
Formula Weight 80.04
Percent Purity >99.0%
Grade Not Available
Packaging Plastic Bottle
Density 1.72 g/3 (20 °C)
Chemical Name or Material Nitric acid ammonia, Ammonia nitrate
Show More Show Less
EINECSNumber 229-347-8
RTECSNumber BR9050000
Recommended Storage +5°C to +30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.