Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Nitrate, Granular, Reagent, ACS, 95%, Spectrum™ Chemical
SDP

Catalog No. 18600631 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
6 x 500 g
12 kg
25 kg
4 x 2.5 kg
50 kg
Packaging:
Poly Bottle
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-631 50 kg Poly Bottle
18-600-624 25 g Poly Bottle
18-600-625 500 g Poly Bottle
18-600-627 2.5 kg Poly Bottle
18-600-626 6 x 500 g Poly Bottle
18-600-628 4 x 2.5 kg Poly Bottle
18-600-629 12 kg Poly Bottle
18-600-630 25 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 18-600-631 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. A121550KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A1215, 6484-52-2, NH4NO3

Spectrum™ Chemical Ammonium Nitrate, Granular, Reagent, ACS is a white crystalline solid which is highly soluble in water. It is most commonly used in agriculture as a nitrogen fertilizer. It is a main component in some explosives and also finds use in instant cold packs as hydrating the salt is an endothermic process. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 6484-52-2
Molecular Formula H4N2O3
Molecular Weight (g/mol) 80.04
MDL Number MFCD00011425
InChI Key DVARTQFDIMZBAA-UHFFFAOYSA-O
IUPAC Name ammonium nitrate
SMILES [NH4+].[O-][N+]([O-])=O

Specifications

pH 4.5-6.0
CAS Max % 100%
Physical Form Granular/Lump
Quantity 50 kg
UN Number UN1942
Percent Purity 95%
Grade Reagent
Packaging Poly Bottle
Residue after Ignition 0.01%
Chemical Name or Material Ammonium Nitrate, Granular
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.