Learn More
Ammonium nitrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Description
It is used in agriculture as a high-nitrogen fertilizer. It is one of the main components of ammonium nitrate/fuel oil (ANFO). ANFO is used in coal mining, quarrying, metal mining, and civil construction. It is also used as an oxidizer in solid rocket propellants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 210°C (decomposition) |
| Melting Point | 170°C |
| Color | Colorless |
| Physical Form | Crystalline |
| Quantity | 250 g |
| Odor | Odorless |
| Appearance | Colorless crystalline |
| Assay Percent Range | 100.00% |
| Density | 1.72 |
| Solubility Information | Highly soluble in water; soluble in alcohol and liquid ammonia. |
| Show More |
Safety and Handling
| Hazard Category | H272-H319 |
| Precautionary Statement | P220-P221-P264b-P271-P280-P280i-P305+P351+P338-P370+P380+P375-P372-P380-P501c |
| DOTInformation | DOT Class: 5.1, Packaging Group: III |
| EINECSNumber | 229-347-8 |
| RTECSNumber | BR9050000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.