Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Nitrate, Purified, Prill, Reagents

Catalog No. 01337495
Change view
Click to view available options
Quantity:
500 g
3 kg
12 kg
25 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-495 12 kg
01-337-494 25 kg
01-337-493 3 kg
01-337-492 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01-337-495 Supplier Reagents Holdings Llc Supplier No. C213550012A5
 Restricted Item
Add to Cart
Add to Cart

Ammonium Nitrate, Purified, Prill, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 6484-52-2
Molecular Formula H4N2O3
Molecular Weight (g/mol) 80.04
MDL Number MFCD00011425
InChI Key DVARTQFDIMZBAA-UHFFFAOYSA-O
Synonym Nitric Acid ammonium salt
IUPAC Name ammonium nitrate
SMILES [NH4+].[O-][N+]([O-])=O

Specifications

Boiling Point 210°C
Melting Point 170°C
Color Colorless/White
Physical Form Solid
Quantity 12 kg
Grade Pure
Packaging Poly Pail
Specific Gravity 1.72
Chemical Name or Material Ammonium Nitrate, Purified, Prill
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.