Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Nitrate, Purified, Spectrum™ Chemical
SDP

Catalog No. 18608844 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-608-844 2.5 kg Poly Pail
18-608-842 500 g Poly Pail
18-608-841 12 kg Poly Pail
18-608-843 50 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-608-844 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. A12172.5KG
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 6484-52-2 | Weight: 80.04 g/mol

Chemical Identifiers

CAS 6484-52-2

Specifications

Physical Form Granular/Lump
CAS Max % 1
Quantity 2.5 kg
Packaging Poly Pail
Molecular Weight (g/mol) 80.04
Grade Purified
Chemical Name or Material Ammonium Nitrate

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.