Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ammonium pentaborate octahydrate

Catalog No. 906034500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-603-4500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-603-4500G Supplier Sigma Aldrich Supplier No. 906034500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ammonium pentaborate is an alkaline salt utilized in the creation of thin films by developing an oxide film which can inhibit corrosion of metals. It finds applications in the preparation of electrolytic capacitors, as a solvent for metallic oxides, as a flame retardant additive, in synthesis of nitrogen-doped ZnO particles, and in treatment of carbon nanotmaterials.

Specifications

CAS 12046-03-6
Quantity 500 g
Linear Formula (NH4)B5O8 -+ 8H2O
Synonym Ammonium boron oxide octahydrate
Percent Purity ≥99.0% (with H2SO4, titration)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.