Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium perchlorate, Reagent Grade, Thermo Scientific Chemicals

Catalog No. AA11658A1
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA11658A1 1 kg
AA1165830 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA11658A1 Supplier Thermo Scientific Chemicals Supplier No. 011658A1
Add to Cart

missing translation for 'validMainframeMsg'

Ammonium perchlorate is used to make rocket propellants, explosives, pyrotechnics, as an etching and engraving agent, and in analytical chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-98-9
Molecular Formula ClH4NO4
Molecular Weight (g/mol) 117.485
MDL Number MFCD00011421
InChI Key HHEFNVCDPLQQTP-UHFFFAOYSA-N
PubChem CID 24639
IUPAC Name azanium;perchlorate
SMILES [NH4+].[O-]Cl(=O)(=O)=O

Specifications

Melting Point (decomposition)
Color White
Physical Form Crystalline
Quantity 1 kg
Solubility Information Freely soluble in water. Soluble in methanol. Slightly soluble in ethanol,acetone. Insoluble in ether,ethyl acetate
Formula Weight 117.5
Grade Reagent
Odor Odorless
Refractive Index 1.482
Appearance White crystalline
Density 1.95
Chemical Name or Material Ammonium perchlorate
Show More Show Less
Hazard Category H201-H271
Precautionary Statement P210-P220-P221-P230a-P240-P250-P280-P283-P306+P360-P370+P378q-P371+P380+P375-P372-P373-P380-P501c
DOTInformation DOT Class: 5.1, Packaging Group: II
EINECSNumber 232-235-1
RTECSNumber SC7520000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.