Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Peroxodisulfate, For Analysis, EMSURE™, ACS, MilliporeSigma™

Catalog No. M1012019012 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
12 kg
Packaging:
Plastic Bucket
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1012019012 12 kg Plastic Bucket
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1012019012 Supplier MilliporeSigma Supplier No. 1012019012
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity grade reagent for analytical and laboratory applications.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7727-54-0
Molecular Formula H8N2O8S2
Molecular Weight (g/mol) 228.19
MDL Number MFCD00003390
InChI Key ROOXNKNUYICQNP-UHFFFAOYSA-N
Synonym ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french
PubChem CID 62648
IUPAC Name diammonium [(sulfonatoperoxy)sulfonyl]oxidanide
SMILES [NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O

Specifications

Melting Point 120°C (decomposition)
pH 3.2 (100g/L, H2O, 20°C)
Physical Form Solid
Quantity 12 kg
Solubility Information 620g/L
Formula Weight 228.19
Percent Purity >98.0%
Grade ACS
Packaging Plastic Bucket
Density 1.98g/3 (20°C)
Chemical Name or Material Ammonium Peroxodisulfate
Show More Show Less
EINECSNumber 231-786-5
RTECSNumber SE0350000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.