Learn More
Ammonium Peroxydisulfate (Crystalline/Certified ACS), Fisher Chemical™
Shop All Fisherbrand ProductsSpecifications
Specifications
| Melting Point | 120°C |
| Color | White |
| pH | 3.2 |
| Physical Form | Powder/Solid |
| Quantity | 10 kg |
| Assay Percent Range | ≥98.0% |
| Linear Formula | (NH4)2S2O8 |
| UN Number | 1444 |
| Formula Weight | 228.19 |
| Percent Purity | ≥98.0% |
| Show More |
Safety and Handling
| Hazard Statement | DANGER! |
| Precautionary Statement | Emergency Overview Oxidizer: Contact with combustible/organic material may cause fire. Causes eye, skin, and respiratory tract irritation. May cause allergic respiratory and skin reaction. Use personal protective equipment. Keep away from clothing and other combustible materials. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. NFPA Health:2 Flammability:1 Instability:2 |
| DOTInformation | DOT Class 5.1, : Oxidizer |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.