Learn More
Strem, An Ascensus Company CAS# 13598-65-7. 25g. Ammonium perrhenate (99+%-Re). MFCD00012377
SDP
Catalog No. 5090113587
Encompass
CAS# 13598-65-7. 25g. Ammonium perrhenate (99+%-Re). MFCD00012377. Molecular Weight: 268.24. Molecular Formula: NH4ReO4. Color/form: white pwdr. Strem# 93-0249. http://www.strem.com/catalog/v/93-0249/
Catalog No. 50-901-13587 Supplier Strem, An Ascensus Company Supplier No. 93024925G
CAS# 13598-65-7. 25g. Ammonium perrhenate (99+%-Re). MFCD00012377. Molecular Weight: 268.24. Molecular Formula: NH4ReO4. Color/form: white pwdr. Strem# 93-0249. http://www.strem.com/catalog/v/93-0249/