Ammonium perrhenate(VII), 99.999% (metals basis), Alfa Aesar™, Puratronic™

Manufacturer: Alfa Aesar 1084609

Quantity 10g

 View more versions of this product

Catalog No. AA1084609

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13598-65-7
    Physical Form Crystalline
    Assay Percent Range 99.999% (Metal basis)
    Molecular Formula NH4ReO4
    Formula Weight 268.24
    MDL Number MFCD00012377
    Chemical Name or Material Ammonium perrhenate(VII)

    Specifications

    Formula Weight 268.24
    Quantity 10g
    Color White
    DOT Information DOT Class: 5.1, Packaging Group: II
    EINECS Number 237-075-6
    Physical Form Crystalline
    Assay Percent Range 99.999% (Metal basis)
    Molecular Formula NH4ReO4
    Appearance White crystalline
    MDL Number MFCD00012377
    CAS 13598-65-7
    Health Hazard 1 H272
    Chemical Name or Material Ammonium perrhenate(VII)

    H272

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit