Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Perrhenate(VII) 99.999%, Thermo Scientific™

Catalog No. AC193850020
Change view
Click to view available options
Quantity:
2 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC193850020 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC193850020 Supplier Thermo Scientific Chemicals Supplier No. 193850020

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13598-65-7
Molecular Formula H4NO4Re
Molecular Weight (g/mol) 268.24
MDL Number MFCD00012377
InChI Key XPSXHXWHJAIQJR-UHFFFAOYSA-M
Synonym azanium oxido trioxo rhenium, ammonium tetraoxorhenate vii, acmc-20alsh, nh4reo4, rhenate reo41-, ammonium, t-4, ammonium tetraoxorhenate 1-, rhenate reo41-, ammonium 1:1 , t-4, ammonium perrhenate, nh4 reo4, rhenate reo41-, ammonium, beta-4, ammonium perrhenate vii , puratronic metals basis
PubChem CID 3084163
IUPAC Name rheniumoylol amine
SMILES N.O[Re](=O)(=O)=O

Specifications

Color White
Physical Form Fine Crystalline Powder
Quantity 2 g
Linear Formula NH4ReO4
Formula Weight 254.24
Density 3.97
Chemical Name or Material Ammonium perrhenate(VII), 99.999%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.