Learn More
Research Products International Corp Ammonium Persulfate [APS], 100 Grams
SDP
Catalog No. 50488481
Encompass_Preferred
Used as a catalyst for acrylamide gel polymerization.
  • Heavy Metals: <0.005%
Catalog No. 50-488-481 Supplier Research Products International Corp Supplier No. A20500100.0
Used as a catalyst for acrylamide gel polymerization.
  • Heavy Metals: <0.005%