Learn More
Sigma Aldrich Fine Chemicals Biosciences Ammonium persulfate, 7727-54-0, MFCD00003390, 100g
Catalog No. NC1336225
Encompass
Linear Formula (NH4)2S2O8, ≥98.0%, Molecular Weight 228.20, ACS reagent, Synonym: AP, APS, Ammonium peroxodisulfate, Ammonium peroxydisulfate, PER; Catalyst for acrylamide gel polymerization.
Catalog No. NC1336225 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 248614100G
Linear Formula (NH4)2S2O8, ≥98.0%, Molecular Weight 228.20, ACS reagent, Synonym: AP, APS, Ammonium peroxodisulfate, Ammonium peroxydisulfate, PER; Catalyst for acrylamide gel polymerization.