Learn More
Sigma Aldrich Fine Chemicals Biosciences Ammonium persulfate, 7727-54-0, MFCD00003390, 25 g
Catalog No. NC1166159
Encompass
Linear Formula: (NH4)2S2O8, Molecular Weight: 228.20, for molecular biology, for electrophoresis, ≥98%, Synonym: AP, APS, Ammonium peroxodisulfate, Ammonium peroxydisulfate, PER. Catalyst for acrylamide gel polymerization. Ammonium persulfate has been used for the preparation of polyacrylamide gels and acrylamide hydrogels.
Catalog No. NC1166159 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A367825G
Linear Formula: (NH4)2S2O8, Molecular Weight: 228.20, for molecular biology, for electrophoresis, ≥98%, Synonym: AP, APS, Ammonium peroxodisulfate, Ammonium peroxydisulfate, PER. Catalyst for acrylamide gel polymerization. Ammonium persulfate has been used for the preparation of polyacrylamide gels and acrylamide hydrogels.