Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Persulfate, 4% (w/w), Reagents

Catalog No. 01336121
Change view
Click to view available options
:
Quantity: 500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-336-121 Quantity: 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-336-121 Supplier Reagents Holdings Llc Supplier No. CA006400500A
Only null left
Add to Cart
Add to Cart

Ammonium Persulfate, 4% (w/w), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7727-54-0
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Ammonium Peroxydisulfate
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Color Colorless
Physical Form Liquid
Quantity 500 mL
Percent Purity 4%
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.0202
Chemical Name or Material Ammonium Persulfate, 4% (w/w)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.