Learn More
Crescent Chemical Co Inc AMMONIUM PERSULFATE
SDP
Supplier:  Crescent Chemical Co Inc SE13375.01
Encompass_Preferred
Ammonium persulfate analytical grade For use in electrophoresis. Polymerisation catalysator. Oxidizing agent of copper, for separation of manganese und chrome. Size - 50G Storage Conditions - +15 °C TO +30 °C Catalog Number - 13375.01 CAS 772
Catalog No. 50-247-286