Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Persulfate, ≥98.0%, ACS Reagent, Solstice

Catalog No. 60045664 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
2.5 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-664 100 g Poly Bottle
60-045-665 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-045-664 Supplier Solstice Supplier No. 248614100G

ACS Reagent,≥98.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7727-54-0
Molecular Formula H8N2O8S2
Molecular Weight (g/mol) 228.19
MDL Number MFCD00003390
InChI Key ROOXNKNUYICQNP-UHFFFAOYSA-N
Synonym ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french
PubChem CID 62648
SMILES [NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O

Specifications

Melting Point 120°C
Color Colorless
Physical Form Solid
Quantity 100 g
Linear Formula (NH4)2S2O8
UN Number UN1444
IUPAC Name diazanium;sulfonatooxy sulfate
Formula Weight 228.2g/mol
Percent Purity ≥98.0%
Grade ACS Reagent
Packaging Poly Bottle
Residue after Ignition ≤0.05%
Density 1.98g/cm3; Vapor Density: 7.9 (vs air)
Chemical Name or Material Ammonium Persulfate
Show More Show Less
Hazard Statement P220-P261-P280-P305 + P351 + P338-P342 + P311
Precautionary Statement H272-H302-H315-H317-H319-H334-H335
DOTInformation 5.1/PGIII
EINECSNumber 231-786-5
RTECSNumber SE0350000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.