Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ammonium persulfate, 98%, extra pure
Click to view available options
Quantity:
100 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent

Specifications
Specifications
Melting Point | 120.0°C |
Color | White to Yellow |
Physical Form | Crystalline Powder and/or Crystals |
Quantity | 100 g |
Assay Percent Range | 98% |
Linear Formula | (NH4)2S2O8 |
Fieser | 01,952; 02,348; 03,238; 05,15; 06,20; 12,33 |
Merck Index | 15, 539 |
Solubility Information | Solubility in water: 582g/L (20°C), may decompose. |
Formula Weight | 228.19 |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause respiratory irritation. Causes skin irritation. May cause an allergic skin reaction. Causes serious eye irritation. |
Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Take any precaution to avoid mixing with combustibles. IF ON SKIN: Wash with plenty of soap and water. If skin irritation or rash occurs. |
EINECSNumber | 231-786-5 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction