Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Persulfate, Molecular Biology Reagent, MP Biomedicals™

Catalog No. 0219398880 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-193-98880 100 g
ICN19398825 25 g
ICN19398890 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02-193-98880 Supplier MP Biomedicals, Inc Supplier No. 0219398880
Add to Cart
Add to Cart

Ammonium Persulfate, Molecular Biology Reagent is a polymerization catalyst used with TEMED for polyacrylamide gel formation.

  • Ammonium persulfate is a polymerization catalyst used with TEMED for polyacrylamide gel formation.
  • APS has also been utilized to study protein-protein interactions via photoinitiated crosslinking chemistry.
  • Other applications of APS include its use as a reducer and retarder in photography, the manufacture of aniline dyes, electroplating, and the decolorizing and deodorizing of oils.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7727-54-0
Molecular Formula H8N2O8S2
Molecular Weight (g/mol) 228.19
MDL Number MFCD00003390
InChI Key ROOXNKNUYICQNP-UHFFFAOYSA-N
Synonym ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french
PubChem CID 62648
SMILES [NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O

Specifications

Melting Point 120°C (Decomposition)
Color White
Physical Form Crystalline Powder
Quantity 100 g
Solubility Information Soluble in water, 83.5 g/100 g water at 25°C
IUPAC Name diammonium [(sulfonatoperoxy)sulfonyl]oxidanide
Formula Weight 228.18
Percent Purity ≥98%
Grade Molecular Biology
Density 1.98 g/cm3 at 68°F; Bulk Density: 951 to 1050 kg/m3
Chemical Name or Material Ammonium persulfate
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.