Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Persulfate, ACS Reagent (Ammonium Peroxydisulfate), Reagents

Catalog No. 01338085
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-338-085 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-338-085 Supplier Reagents Holdings Llc Supplier No. C1136500500A5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ammonium Persulfate, ACS Reagent (Ammonium Peroxydisulfate), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7727-54-0
Molecular Formula H8N2O8S2
Molecular Weight (g/mol) 228.19
InChI Key ROOXNKNUYICQNP-UHFFFAOYSA-N
Synonym Ammonium Peroxydisulfate
IUPAC Name diammonium [(sulfonatoperoxy)sulfonyl]oxidanide
SMILES [NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O

Specifications

Melting Point 120°C to 160°C
Color Colorless/White
Physical Form Solid
Quantity 500 g
Grade ACS
Packaging Poly Bottle
Specific Gravity 1.98
Chemical Name or Material Ammonium Persulfate, ACS Reagent (Ammonium Peroxydisulfate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.