Learn More
Research Products International Corp Ammonium Persulfate [APS], 10 Grams
SDP
Catalog No. 501368040
Encompass_Preferred
Used as a catalyst for acrylamide gel polymerization.
  • Heavy Metals: <0.005%
Catalog No. 50-136-8040 Supplier Research Products International Corp Supplier No. A2050010.0
Used as a catalyst for acrylamide gel polymerization.
  • Heavy Metals: <0.005%