Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALADDIN SCIENTIFIC CORPORATION Ammonium phosphate monobasic, 100g

Catalog No. 5000082495
Encompass_Preferred

Ammonium phosphate monobasic Chromatographic grade 99%(T)Cas-7722-76-1

Catalog No. 50-000-82495 Supplier ALADDIN SCIENTIFIC CORPORATION Supplier No. A111775100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ammonium phosphate monobasic Chromatographic grade 99%(T)Cas-7722-76-1

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.