Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Phosphate Monobasic, ACS Reagent (Ammonium Dihydrogen Phosphate), Reagents

Catalog No. 01338084
Change view
Click to view available options
:
Quantity: 500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-338-084 Quantity: 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-338-084 Supplier Reagents Holdings Llc Supplier No. C1137000500C2
Only null left
Add to Cart
Add to Cart

Ammonium Phosphate Monobasic, ACS Reagent (Ammonium Dihydrogen Phosphate), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7722-76-1
Molecular Formula H6NO4P
Molecular Weight (g/mol) 115.03
MDL Number MFCD00003396
InChI Key LFVGISIMTYGQHF-UHFFFAOYSA-N
Synonym Ammonium dihydrogen phosphate, Monoammonium phosphate
IUPAC Name phosphoric acid amine
SMILES N.OP(O)(O)=O

Specifications

Boiling Point 197°C
Melting Point 190°C
Color Colorless/White
Physical Form Solid
Quantity 500 g
Grade ACS
Packaging Glass Bottle
Specific Gravity 1.803
Chemical Name or Material Ammonium Phosphate Monobasic, ACS Reagent (Ammonium Dihydrogen Phosphate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.