Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Phosphate Solution, 400 g (NH4)2HPO4 + 800 mL Water, Ricca Chemical
SDP

Catalog No. R06900004A Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R06900004A 4 L Natural Poly Bottle
6904 120 mL Natural Poly Bottle
690-16 500 mL Natural Poly Bottle
R06900001A 1 L Natural Poly Bottle
R069000020F 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. R06900004A Supplier Ricca Chemical Company Supplier No. 6901
Add to Cart
Add to Cart

APHA, EPA for Sulfide Testing

Chemical Identifiers

Molecular Formula H9N2O4P
Molecular Weight (g/mol) 132.06
MDL Number MFCD00010891
InChI Key MNNHAPBLZZVQHP-UHFFFAOYSA-N
Synonym diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate
PubChem CID 24540
ChEBI CHEBI:63051
IUPAC Name phosphoric acid diamine
SMILES N.N.OP(O)(O)=O

Specifications

Color Colorless
CAS 7783-28-0, 7732-18-5
CAS Min % 65.0, 32.68
CAS Max % 68.0, 34.01
Grade Laboratory
Packaging Natural Poly Bottle
pH ∼8
Quantity 4 L
Physical Form Liquid
Concentration or Composition Notes 400 g (NH4)2HPO4 + 800mL Water
Chemical Name or Material Ammonium Phosphate Solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.