Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Phosphate Solution for Sulfide 400 g Ammonium Phosphate + 800 mL Water, Reagents

Catalog No. 01336006
Change view
Click to view available options
:
Quantity: 500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-336-006 Quantity: 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-336-006 Supplier Reagents Holdings Llc Supplier No. CA016700500A
Only null left
Add to Cart
Add to Cart

Ammonium Phosphate Solution for Sulfide 400 g Ammonium Phosphate + 800 mL Water, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Ammonium Phosphate Solution for Sulfide 400 g Ammonium Phosphate and 800 mL Water
Boiling Point 106°C
Color Colorless
CAS 7732-18-5, 7783-28-0
pH 8
Physical Form Liquid
Quantity 500 mL
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.1481
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.