Learn More
Strem, An Ascensus Company CAS# 54723-94-3. 50g. Ammonium phosphomolybdate hydrate, reagent. MFCD00150423
SDP
Catalog No. 5090113583
Encompass
CAS# 54723-94-3. 50g. Ammonium phosphomolybdate hydrate, reagent. MFCD00150423. Molecular Weight: 1876.35. Molecular Formula: (NH4)3PO4∙12MoO3∙XH2O. Color/form: yellow pwdr. Strem# 93-0248. http://www.strem.com/catalog/v/93-0248/
Catalog No. 50-901-13583 Supplier Strem, An Ascensus Company Supplier No. 93024850G
CAS# 54723-94-3. 50g. Ammonium phosphomolybdate hydrate, reagent. MFCD00150423. Molecular Weight: 1876.35. Molecular Formula: (NH4)3PO4∙12MoO3∙XH2O. Color/form: yellow pwdr. Strem# 93-0248. http://www.strem.com/catalog/v/93-0248/