Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ammonium phosphomolybdate hydrate

Catalog No. 34216550G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-216-550G 50 g
34-216-5250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 34-216-550G Supplier Sigma Aldrich Supplier No. 34216550G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ammonium phosphomolybdate hydrate

Specifications

CAS 54723-94-3
Molecular Formula H12Mo12N3O40P -+ xH2O
Linear Formula (NH4)3 PMo12O40 -+ xH2O
Quantity 50 g
MDL Number MFCD00150423
Synonym Ammonium-12-molybdophosphate; Molybdophosphoric acid ammonium salt; Phosphomolybdic acid ammonium salt hydrate
Molecular Weight (g/mol) 1876.35
Percent Purity ≥61.1% (%Mo, gravimetric)
Biological Activity Respiratory system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.