Learn More
Hach Company Intellical™ ISENH4181 Ammonium (NH₄⁺) Ion Selective Electrode (ISE), 3 m cable
Catalog No. 501997731 Shop All Hach Company Products
Encompass
Intellical™ ISENH4181 is a digital, combination, ammonium ion selective electrode (ISE) with a with a non-refillable gel Dritek reference with a double junction, ceramic porous pin and porous PTFE annular ring, and built-in temperature sensor. The electrode measures ammonium concentration in water samples. The laboratory version of this ISE is shockproff with its Epoxy plastic body.The Intellical ISENH4181 probe is available with a 1 or 3 meter cable and is intended for laboratory use. The ISENH4181 is ideal for measuring ammonium concentrations in wastewater, drinking water and general water quality applications.
Catalog No. 50-199-7731 Supplier Hach Company Supplier No. ISENH418103
Add to Cart
Add to Cart
Intellical™ ISENH4181 is a digital, combination, ammonium ion selective electrode (ISE) with a with a non-refillable gel Dritek reference with a double junction, ceramic porous pin and porous PTFE annular ring, and built-in temperature sensor. The electrode measures ammonium concentration in water samples. The laboratory version of this ISE is shockproff with its Epoxy plastic body.The Intellical ISENH4181 probe is available with a 1 or 3 meter cable and is intended for laboratory use. The ISENH4181 is ideal for measuring ammonium concentrations in wastewater, drinking water and general water quality applications.