Learn More
Medchemexpress LLC Ammonium purpurate | 3051-09-0 | MFCD00012777 | 98.0% | 284.19 g/mol | C8H8N6O6 | 100G
SDP
Catalog No. 5000234994
Encompass_Preferred
Murexide (ammonium purpurate) is an analytical reagent supplied as a solid powder used primarily as an indicator in complexometric titrations, for example to determine water hardness. It is a brown to purplish red solid (molecular weight 284.19 g/mol, formula C8H8N6O6) with a reported purity of 98.0%, commonly offered in 100 g packs for laboratory use.
  • Used as an indicator in complexometric titrations to detect metal ions.
  • High purity (98.0%) suitable for analytical applications.
  • Solid powder form simplifies storage and handling.
  • Distinct brown to purplish red color for easy identification.
  • Molecular weight 284.19 g/mol and formula C8H8N6O6 for reference.
  • Available in 100 g laboratory pack sizes.
Catalog No. 50-002-34994 Supplier Medchemexpress LLC Supplier No. HY125929100G
Murexide (ammonium purpurate) is an analytical reagent supplied as a solid powder used primarily as an indicator in complexometric titrations, for example to determine water hardness. It is a brown to purplish red solid (molecular weight 284.19 g/mol, formula C8H8N6O6) with a reported purity of 98.0%, commonly offered in 100 g packs for laboratory use.
  • Used as an indicator in complexometric titrations to detect metal ions.
  • High purity (98.0%) suitable for analytical applications.
  • Solid powder form simplifies storage and handling.
  • Distinct brown to purplish red color for easy identification.
  • Molecular weight 284.19 g/mol and formula C8H8N6O6 for reference.
  • Available in 100 g laboratory pack sizes.