Learn More
eMolecules​ 5108-96-3 | Ammonium pyrrolidine-1-carbodithioate | Oakwood Chemicals | MFCD00012720 | 164.290 | C5H12N2S2 | 95.000 | N.SC(=S)N1CCCC1 | 25g | 480163968
Catalog No. 502148456 Shop All eMolecules Inc. Products
Encompass_Preferred
Ammonium pyrrolidine-1-carbodithioate | Oakwood Chemicals | 5108-96-3 | MFCD00012720 | 164.290 | C5H12N2S2 | 95.000 | N.SC(=S)N1CCCC1 | 25g | 480163968
Catalog No. 50-214-8456 Supplier eMolecules​ Supplier No. 480163968
Ammonium pyrrolidine-1-carbodithioate | Oakwood Chemicals | 5108-96-3 | MFCD00012720 | 164.290 | C5H12N2S2 | 95.000 | N.SC(=S)N1CCCC1 | 25g | 480163968