Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ammonium Pyrrolidinedithiocarbamate

Catalog No. 0993525G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-935-25G 25 g
09-935-1KG 1 kg
09-935-100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 09-935-25G Supplier Sigma Aldrich Supplier No. 0993525G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ammonium Pyrrolidinedithiocarbamate

Specifications

CAS 5108-96-3
Melting Point 153°C to 155°C
Molecular Formula C5H12N2S2
Linear Formula C5H9NS2 -+ NH3
MDL Number MFCD00012720
Quantity 25 g
Synonym 1-Pyrrolidinecarbodithioic acid ammonium salt; APDC; Ammonium pyrrolidinecarbodithioate; PDC; PDTC
Molecular Weight (g/mol) 164.29 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.