Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ammonium sodium phosphate dibasic tetrahydrate

Catalog No. 503125814 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-312-5814 1 kg
50-312-5613 100 g
50-312-5610 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-312-5814 Supplier Sigma Aldrich Supplier No. S95061KG
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ammonium sodium phosphate dibasic tetrahydrate

Specifications

CAS 7783-13-3
Molecular Formula H5NNaO4P -+ 4H2O
Linear Formula NaNH4HPO4 -+ 4H2O
MDL Number MFCD00066970
Quantity 1 kg
Synonym sec-Sodium ammonium phosphate; Ammonium sodium hydrogen phosphate; Sodium ammonium hydrogen phosphate
Molecular Weight (g/mol) 209.07
Percent Purity ≥99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.