Learn More
Hach Company Ammonium Stand Sol 2.5mg/L
Supplier:  Hach Company LCA505
Encompass
Ammonium Standard Solution with 2 5 mg/L NH -N for quality control checks with LCK505 Ammonium cuvette test LCA505 is also suitable for quality checks of the NH6000sc and the Amtax sc ammonia analyzers
Catalog No. 50-002-06978