Learn More
Hach Company Ammonium Stand Sol 50 mg/L
Catalog No. 5000207038 Shop All Hach Company Products
Encompass
Ammonia Standard Solution with 50 mg/L NH -N for quality control checks with TNT837 Ammonia vial test LCA503 is also suitable for quality checks with the Amtax sc Ammonium analyzer
Catalog No. 50-002-07038 Supplier Hach Company Supplier No. LCA503
Ammonia Standard Solution with 50 mg/L NH -N for quality control checks with TNT837 Ammonia vial test LCA503 is also suitable for quality checks with the Amtax sc Ammonium analyzer