Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ammonium sulfamate | 7773-06-0 | 99.7% | 114.12 g/mol | H6N2O3S | 100 G
SDP

Catalog No. 5000209101
Encompass_Preferred

Ammonium sulfamate is an inorganic ammonium salt (sulfamic acid ammonium) used as a reagent in biochemical and life science research. It is supplied as a high-purity crystalline powder and intended for research use only. Safety data sheets are available for handling and storage information.

  • High assay purity: 99.7%.
  • Cas number: 7773-06-0.
  • Chemical formula: H6N2O3S; molar mass ~114.12 g/mol.
  • Available in common lab pack sizes, including 100 g.
  • Supplied for research use; consult SDS for safety and handling.

Catalog No. 50-002-09101 Supplier Medchemexpress LLC Supplier No. HYW040155100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ammonium sulfamate is an inorganic ammonium salt (sulfamic acid ammonium) used as a reagent in biochemical and life science research. It is supplied as a high-purity crystalline powder and intended for research use only. Safety data sheets are available for handling and storage information.

  • High assay purity: 99.7%.
  • Cas number: 7773-06-0.
  • Chemical formula: H6N2O3S; molar mass ~114.12 g/mol.
  • Available in common lab pack sizes, including 100 g.
  • Supplied for research use; consult SDS for safety and handling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.