Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ammonium sulfamate | 7773-06-0 | MFCD00011429 | 99.7% | 114.12 | H6N2O3S | 1 KG
SDP

Catalog No. 5000208771
Encompass_Preferred

Ammonium sulfamate is an inorganic salt used as a laboratory reagent and specialty chemical in life-science research and related applications. It is supplied as a white to off-white solid and is commonly used for biochemical assays, sample preparation, and other laboratory processes.

  • Purity 99.65%.
  • CAS number 7773-06-0.
  • Molecular weight 114.12 g/mol.
  • Chemical formula H6N2O3S.
  • Appearance white to off-white solid.
  • Typical applications biochemical research and laboratory reagent use.
  • Available in 1 kg package.

Catalog No. 50-002-08771 Supplier Medchemexpress LLC Supplier No. HYW0401551KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ammonium sulfamate is an inorganic salt used as a laboratory reagent and specialty chemical in life-science research and related applications. It is supplied as a white to off-white solid and is commonly used for biochemical assays, sample preparation, and other laboratory processes.

  • Purity 99.65%.
  • CAS number 7773-06-0.
  • Molecular weight 114.12 g/mol.
  • Chemical formula H6N2O3S.
  • Appearance white to off-white solid.
  • Typical applications biochemical research and laboratory reagent use.
  • Available in 1 kg package.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.