Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Fine Chemicals Biosciences AMMONIUM SULFAMATE, 98%

Catalog No. NC1803029
Encompass

Ammonium sulfamate | Purity: 98% | 7773-06-0 | MFCD00011429 | 100 g

Catalog No. NC1803029 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 228745100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Ammonium sulfamate | Purity: 98% | 7773-06-0 | MFCD00011429 | 100 g

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.