Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Sulfamate, Puriss. p.a., ≥99.0% (T), Solstice

Catalog No. 60044628 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Poly bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-044-628 100 g Poly bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-044-628 Supplier Solstice Supplier No. 09960100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Puriss. p.a.,≥99.0% (T)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7773-06-0
Molecular Formula H6N2O3S
Molecular Weight (g/mol) 114.119
MDL Number MFCD00011429
InChI Key GEHMBYLTCISYNY-UHFFFAOYSA-N
Synonym ammonium sulfamate, ammonium sulphamate, ammate, ikurin, ammonium amidosulfonate, feliderm k, ammate x, sulfamic acid, monoammonium salt, ammonium sulfamidate, ammonium sulphamidate
PubChem CID 24482
ChEBI CHEBI:81950
IUPAC Name azanium;sulfamate
SMILES [NH4+].NS(=O)(=O)[O-]

Specifications

Melting Point 131°C to 135°C
Color White
pH 4.5 to 6.0 (at 114.1g/L 25°C)
Physical Form Crystalline
Quantity 100 g
Linear Formula NH4SO3NH2
UN Number UN3077
Formula Weight 114.12g/mol
Percent Purity ≥99.0% (T)
Grade puriss. p.a.
Packaging Poly bottle
Residue after Ignition ≤0.02% (as SO4)
Chemical Name or Material Ammonium Sulfamate
Show More Show Less
Hazard Statement P273
Precautionary Statement H302-H400
DOTInformation 9/PGIII
EINECSNumber 231-871-7
RTECSNumber WO6125000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.