Learn More
Research Products International Corp Ammonium Sulfate, 1 Kilogram
SDP
Catalog No. 50488483
Encompass_Preferred
Useful for precipitation or fractionation of proteins or for purification of antibodies. Used in crystallographic analysis of nucleic acids and proteins.
Catalog No. 50-488-483 Supplier Research Products International Corp Supplier No. A205101000.0
Useful for precipitation or fractionation of proteins or for purification of antibodies. Used in crystallographic analysis of nucleic acids and proteins.