Learn More
Biotang Inc AMMONIUM SULFATE 1M, 50 ML, MT-125,1 M Ammonium Sulfate solution ((NH4)2SO4). Prepared in 18.2 megohms water and filtered through 0.22-micron filter. Shipped at room temperature. Store at room temperature.
Catalog No. 50152264 Shop All BIOTANG Inc Products
Encompass_Preferred
AMMONIUM SULFATE 1M, 50 ML, MT-125,1 M Ammonium Sulfate solution ((NH4)2SO4). Prepared in 18.2 megohms water and filtered through 0.22-micron filter. Shipped at room temperature. Store at room temperature.
Catalog No. 50-152-264 Supplier Biotang Inc Supplier No. BTBB811
AMMONIUM SULFATE 1M, 50 ML, MT-125,1 M Ammonium Sulfate solution ((NH4)2SO4). Prepared in 18.2 megohms water and filtered through 0.22-micron filter. Shipped at room temperature. Store at room temperature.