Learn More
Research Products International Corp Ammonium Sulfate, 25 KG
SDP
Catalog No. 501977166
Encompass_Preferred
Useful for precipitation or fractionation of proteins or for purification of antibodies. Used in crystallographic analysis of nucleic acids and proteins.
Catalog No. 50-197-7166 Supplier Research Products International Corp Supplier No. A2051025000.0
Useful for precipitation or fractionation of proteins or for purification of antibodies. Used in crystallographic analysis of nucleic acids and proteins.